missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin A Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99327APC
This item is not returnable.
View return policy
Description
Cystatin A Polyclonal antibody specifically detects Cystatin A in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Cystatin A | |
| Polyclonal | |
| ELISA, Immunohistochemistry-Paraffin | |
| cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
| Produced in rabbits immunized with purified, recombinant Human Cystatin A (Uniprot#: P01040; Ile 2-Phe 98) | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 1475 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction