missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin C Recombinant Rabbit Monoclonal Antibody (23GB1845), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00€ - 562.00€
Specifications
| Antigen | Cystatin C |
|---|---|
| Clone | 23GB1845 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089938
![]() |
Invitrogen™
MA553742 |
100 μL |
562.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088362
|
Invitrogen™
MA553743 |
20 μL |
207.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin C Recombinant Monoclonal antibody specifically detects Cystatin C in Human, Mouse samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| Cystatin C | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P01034, P21460 | |
| 13010, 1471 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1845 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227 | |
| A synthesized peptide derived from human Cystatin C (1-146AA). | |
| Primary | |
| -20°C | |
| Cst3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title