missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin/Stefin Homolog Antibody (RM0218-8D33) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12412-0.025MG
This item is not returnable.
View return policy
Description
Cystatin/Stefin Homolog Monoclonal specifically detects Cystatin/Stefin Homolog in Mouse samples. It is validated for Western Blot, Block/Neutralize.
Specifications
| Cystatin/Stefin Homolog | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:100-1:2000, Blocking/Neutralizing 50-200 molar excess | |
| XP_148650 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 1475 | |
| Mouse | |
| Purified |
| Blocking Assay, Western Blot | |
| RM0218-8D33 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CSTA | |
| Mouse recombinant Cystatin H (Cystatin/Stefin Homolog) | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction