missing translation for 'onlineSavingsMsg'
Learn More

Cytisine, TRC

Product Code. 18023050
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18023050 10 mg 10mg
18071840 500 mg 500mg
18064030 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18023050 Supplier Toronto Research Chemicals Supplier No. TRCC99850010MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-35-8
Molecular Formula C11 H14 N2 O
Molecular Weight (g/mol) 190.24
Synonym (-)-Cytisine, (1R,5S)-1,2,3,4,5,6-Hexahydro-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, 1,2,3,4,5,6-hexahydro-, (1R)-, Cytisine (6CI,8CI), (1R,5S)-1,2,3,4,5,6-Hexahydro-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, (-)-Cytisine, Baptitoxin, Baptitoxine, Cytisin, Cytiton, Cytitone, Laburnin, Sophorin, Sophorine, Tabex, Tsitafat, Ulexin, Ulexine
IUPAC Name (1R,9S)-7,11-diazatricyclo[7.3.1.02,7]trideca-2,4-dien-6-one
SMILES O=C1C=CC=C2[C@H]3CNC[C@H](C3)CN12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H14N2O/c14-11-3-1-2-10-9-4-8(5-12-6-9)7-13(10)11/h1-3,8-9,12H,4-7H2/t8-,9+/m0/s1
Formula Weight 190.1106
Percent Purity >95%
Chemical Name or Material Cytisine (Laburnin)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.