missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome p450 2J2 Antibody (OTI5C5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70539G
This item is not returnable.
View return policy
Description
Cytochrome p450 2J2 Monoclonal specifically detects Cytochrome p450 2J2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cytochrome p450 2J2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP2J2 | |
| Full length recombinant protein of human Cytochrome p450 2J2 (NP_000766) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI5C5 | |
| Western Blot, Immunohistochemistry | |
| Arachidonic acid epoxygenase, CPJ2, CYPIIJ2, cytochrome P450 2J2, cytochrome P450, family 2, subfamily J, polypeptide 2, cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 1573.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction