missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 4A Antibody (M25-P2A10), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50212FR
This item is not returnable.
View return policy
Description
Cytochrome P450 4A Monoclonal specifically detects Cytochrome P450 4A in Human, Mouse, Rat, Drosophila, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Cytochrome P450 4A | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP4A11 | |
| Peptide Sequence - KNGIHLRLR (amino acids 499 - 507) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1579 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| M25-P2A10 | |
| Western Blot, ELISA, Immunohistochemistry | |
| 20-HETE synthase, CP4Y, CYP4A2, cytochrome P450 4A11, cytochrome P450, family 4, subfamily A, polypeptide 11, cytochrome P450, subfamily IVA, polypeptide 11, cytochrome P-450HK-omega, cytochrome P450HL-omega, fatty acid omega-hydroxylase, lauric acid omega-hydroxylase, P450HL-omega | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Drosophila, Xenopus species | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu