missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 4A Antibody (M25-P2A10), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50212JF549
This item is not returnable.
View return policy
Description
Cytochrome P450 4A Monoclonal specifically detects Cytochrome P450 4A in Human, Mouse, Rat, Drosophila, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| CytochromeP4504A | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP4A11 | |
| Peptide Sequence - KNGIHLRLR (amino acids 499 - 507) | |
| 0.1 mL | |
| Primary | |
| Drosophila, Human, Mouse, Rat, Xenopus species | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| M25-P2A10 | |
| Western Blot, ELISA, Immunohistochemistry | |
| 20-HETE synthase, CP4Y, CYP4A2, cytochrome P450 4A11, cytochrome P450, family 4, subfamily A, polypeptide 11, cytochrome P450, subfamily IVA, polypeptide 11, cytochrome P-450HK-omega, cytochrome P450HL-omega, fatty acid omega-hydroxylase, lauric acid omega-hydroxylase, P450HL-omega | |
| Mouse | |
| Protein A purified | |
| Lipid and Metabolism | |
| 1579 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction