Learn More
BTX™ CytoGo™ Bioburden Tested Electroporation Cuvette
Description
With BTX Cuvettes Plus CytoGo traceable electroporation cuvettes, you can advance therapeutic research and development and production with confidence, ensuring consistency, compliance, and reproducibility at every stage.
- Available in 1 mm, 2 mm, and 4 mm gap formats to support controlled and reproducible electroporation conditions across a broad range of cell-therapy-relevant cell types
- Designed to support electric field strengths of up to 25 kV/cm
- Each cuvette is sterile, single-use, and cytotoxin- and endotoxin-tested
- A QR code on each cuvette provides traceability of every electroporation experiment
Available as a 50-pack, the Cuvettes Plus CytoGo™ cuvettes provide full traceability of raw materials, manufacturing and sterilization lots, and are tested for low cytotoxicity and endotoxin levels–supporting reliable scale-up from development through production
Specifications
Specifications
| Sterility | Sterile |
| Type | Electroporation Cuvette |
| Disposable | Single-use |
| Capacity (Metric) | 20 to 90 μL |
| Material | Polycarbonate, Polished Aluminum |
| Quantity | 50 Pk. |
| Width (Metric) Gap | 1 mm |
| For Use With (Equipment) | Electroporator |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |