missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 2 Antibody (10A12), Novus Biologicals™
Mouse Monoclonal Antibody
195.00€
Specifications
| Antigen | Cytohesin 2 |
|---|---|
| Clone | 10A12 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 3 - 5 μg/ 106 cells, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:25 - 1:200, Western Blot (Negative) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18773114
|
Novus Biologicals
NBP2-22514-0.025mg |
0.025 mg | |||||||
Description
Cytohesin 2 Monoclonal specifically detects Cytohesin 2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
| Cytohesin 2 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| Q99418 | |
| 9266 | |
| Purified, His-tagged cytohesin-2. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 10A12 | |
| Flow Cytometry 3 - 5 μg/ 106 cells, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:25 - 1:200, Western Blot (Negative) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ARF exchange factor, ARF nucleotide-binding site opener, ARNOPH, SEC7 and coiled-coil domain-containing protein 2, CTS18.1, cytohesin 2, pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2), Protein ARNO, PSCD2LCTS18, PSCD2pleckstrin homology, Sec7 and coiled-coil domains 2, Sec7 and coiled/coil domains 2 (cytohesin-2) | |
| CYTH2 | |
| IgG1 | |
| PEG purified | |
| This antibody reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title