Learn More
Cytokeratin 14 Antibody (CL14568) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26869CL1
Product Code. 30301168
  • 1210.00€ / 0.10mL
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 14
Monoclonal
CoraFluor 1
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ
This antibody was developed using a synthetic peptide derived from P02533, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers
3861
Store at 4⪚C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
CL14568