missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 14 Antibody (KRT14/4131), Novus Biologicals™
Mouse Monoclonal Antibody
249.00€ - 494.00€
Specifications
| Antigen | Cytokeratin 14 |
|---|---|
| Clone | KRT14/4131 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607144
|
Novus Biologicals
NBP3-20245-100ug |
100 μg |
494.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696154
|
Novus Biologicals
NBP3-20245-20ug |
20 μg |
249.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 14 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10mM PBS with 0.05% BSA | |
| 3861 | |
| IgG1 κ | |
| Protein A or G purified |
| KRT14/4131 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
| Recombinant fragment (around aa351-472) of human Cytokeratin 14 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title