missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534835
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, NT2D1. Predicted reactivity: Mouse (85%), Rat (87%), Dog (87%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21. 2.
Specifications
| Cytokeratin 15 | |
| Polyclonal | |
| Unconjugated | |
| KRT15 | |
| AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3866 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.98 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P19012 | |
| KRT15 | |
| Recombinant protein encompassing a sequence within the center region of human Cytokeratin 15. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction