missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Polyclonal Antibody
Cytokeratin 19 Polyclonal Antibody
Brand: Invitrogen PA5141116
This item is not returnable.
View return policy
Description
Cytokeratin 19 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| Cytokeratin 19 | |
| Polyclonal | |
| Unconjugated | |
| KRT19 | |
| 40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16669, 360626, 3880 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P08727, P19001, Q63279 | |
| KRT19 | |
| Recombinant protein within Human Cytokeratin 19 aa 100-350. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction