missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
113.00€ - 363.00€
Specifications
| Antigen | Cytokeratin 2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640684
|
Novus Biologicals
NBP3-21424-25ul |
25 μg |
113.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633684
|
Novus Biologicals
NBP3-21424-100ul |
100 μg |
363.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 2 Polyclonal antibody specifically detects Cytokeratin 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS, pH 7.0 | |
| 3849 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CK-2e, Cytokeratin-2e, epidermal ichthyosis bullosa of Siemens, Epithelial keratin-2e, K2e, keratin 2, keratin 2A (epidermal ichthyosis bullosa of Siemens), keratin, type II cytoskeletal 2 epidermal, Keratin-2 epidermis, keratin-2e, KRT2AMGC116967, KRT2EMGC116968, KRTE, Type-II keratin Kb2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Cytokeratin 2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title