missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 20 Antibody (CL9390) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88932AF700
This item is not returnable.
View return policy
Description
Cytokeratin 20 Monoclonal antibody specifically detects Cytokeratin 20 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 20 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CD20, CK20, CK-20, cytokeratin-20, K20cytokeratin 20, keratin 20, keratin, type I cytoskeletal 20, keratin-20, KRT21, MGC35423, Protein IT | |
| This antibody was developed using a recombinant protein derived from P35900, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 54474 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL9390 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction