missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 20 Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody
185.00€ - 415.00€
Specifications
| Antigen | Cytokeratin 20 |
|---|---|
| Clone | C2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17979582
![]() |
Invitrogen™
MA551149 |
200 μL |
415.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17969182
|
Invitrogen™
MA551150 |
20 μL |
185.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.Specifications
| Cytokeratin 20 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
| KRT20 | |
| IgG1 κ | |
| Protein A/G | |
| KRT20 |
| C2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| P35900, Q29218 | |
| 100520330, 54474 | |
| Recombinant Human CK 20 protein, His9-Ile424 (Accession #P35900). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title