missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 20 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
129.00€ - 399.00€
Specifications
| Antigen | Cytokeratin 20 |
|---|---|
| Clone | 2H10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236428
|
Proteintech
82428-1-RR-100UL |
100 μL |
439.00€ 399.00€ / 100µL Save 40.00€ 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236321
|
Proteintech
82428-1-RR-20UL |
20 μL |
149.00€ 129.00€ / 20µL Save 20.00€ 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Cytokeratin 20 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Cytokeratin 20. This antibody recognizes human, mouse, rat, pig antigen. The Cytokeratin 20 antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.Specifications
| Cytokeratin 20 | |
| 1000 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 54474 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2H10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Pig | |
| CD20; CK20; K20; KRT21; MGC35423 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title