missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 39 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00390792-B01P-50ug
This item is not returnable.
View return policy
Description
Cytokeratin 39 Polyclonal antibody specifically detects Cytokeratin 39 in Human samples. It is validated for Western Blot
Specifications
| Cytokeratin 39 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| CK-39, Cytokeratin-39, K39, KA35, keratin 39, Keratin, Type I Cytoskeletal 39, Keratin-39, KRT39, Type I Hair Keratin Ka35 | |
| KRT39 (AAI60073.1, 1 a.a. - 491 a.a.) full-length human protein. MDTKGCTTTNSPSTPCQNCSRITNVSTISSNNGCHPGGLTVNNCQPAGHVLRIPWDQGCQPTPRFCRKPIYLMNNFNARFSLDDCSWYGEGINSNEKETMQILNERLANYLQKVRMLERENAELESKIQEESNKELPVLCPDYLSYYTTIEELQQKILCTKAENSRLVSQIDNTKLTADDLRAKYEAEVSLRQLVESDANGLKQILNVLTLGKADLEAQVQSLKEELLCLKNNHKEEINSLQCQLGERLDIEVTAAPSADLNQVLQEMRCQYEPIMETNRKDVEQWFNTQIEELNQQVVTSSQQQQCCQKEIIELRRSVNTLEVELQAQHRMRDSQECILTETEARYTALLTQIQSLIDNLEAQLAEIRCALERQNQEYEILLDVKSRLECEITTYRSLLESSDGKRPCYPRATKCEPSPWTSCKSGAIESTAPACTSSSPCSLKEHCSACGPLSRILVKICTITKEIKDGKVISSYEHVQPCFIIRPAKV | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAI60073.1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 390792 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction