missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8/18 Mouse, Clone: KRT8.18/1346, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 478.00€
Specifications
| Antigen | Cytokeratin 8/18 |
|---|---|
| Clone | KRT8.18/1346 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15911408
|
EnquireBio
MSM27-1346-P0 |
20 μg | |||||||
|
15921408
|
EnquireBio
MSM27-1346-P1 |
100 μg |
554.40€ 478.00€ / 100µg Save 76.40€ 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Cytokeratin 8/18 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic |
| KRT8.18/1346 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Cytoskeleton preparation from HeLa cells | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| MCF-7 or A431 cells. Skin, Colon, lung or breast carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title