missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17514-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specifically against KRT8.
Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.Specifications
| Cytokeratin 8 | |
| Polyclonal | |
| Unconjugated | |
| KRT8 | |
| CARD2, CK 8, CK8, CYK8, Cytokeratin 8, K2C8, K8, keratin 8, KO, KRT8, Type II keratin Kb8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25626, 3856 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05787, Q10758 | |
| KRT8 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction