missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACE/ADAM17 Antibody (MM0561-8C13), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12018UV
This item is not returnable.
View return policy
Description
TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TACE/ADAM17 | |
| Monoclonal | |
| DyLight 350 | |
| P78536 | |
| ADAM17 | |
| Human recombinant TACE/ADAM17 | |
| 0.1 mL | |
| Apoptosis, Stem Cell Signaling Pathway | |
| 6868 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| MM0561-8C13 | |
| Western Blot, Flow Cytometry | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction