missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cytomat™ 10 C450 Series Automated Incubators

Product Code. 18979521 Shop All Thermo Scientific Products
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
18979521 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 18979521 Supplier Thermo Scientific™ Supplier No. 51031168
 Restricted Item
Estimated Shipment: 02-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Temperature Range: 4°C to 50°C with External Chiller; Relative Humidity: <98% at 37°C

Designed to fit into the busiest of cell biology or screening laboratories, Thermo Scientific™ Cytomat™ 10 C450 wide temperature range incubator and storage unit is the ideal solution for high capacity incubation (210 SBS microplates) of primary cell or cell line cultures with a wide temperature application range combined with 4°C storage. The latest product of the Cytomat 10 series can be seamless integrated with existing automated assay hardware and provides mean access time below 10 seconds combined with controlled humidity and thermal decontamination routine ContraCon.

Matches your Capacity with 10 Stacker
  • 280pcs. 1536-well plates
  • 210pcs. 96-well or 384-well plates
  • 150pcs. transwell or insert plates
  • 100pcs. deep-well

â–oFurther stacker types on request


Effortless Humidity Control
  • HydraSmart Technology - Ultrasonic vapor technology
  • Sterilized water supply with external tank

Fast
  • Below 10sec. mean access time

Contamination Prevention
  • Automated decontamination routine - ContraCon
  • Solid copper inner chamber (option)
  • H2O2-flange (option)

Convenient – Easy to use
  • 5” Touch screen as standard
  • Internal and direct barcode reader
  • Various gate / transfer positions

Many further options
  • NextGen Display (NGD)
  • CyMon - Cytomat Remote Monitioring Application
  • O2 control
  • high Humidity (hH) up to 98% r.H. at 37°C
  • SILA (Standard in Lab Automation) compliant

Certification
  • CE
  • RoHS

Manufactured in ISO 9001 and ISO 13485 accredited facility

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Access Time Below 10 sec.
Chamber Material Copper
Holds 280 Pc. 1536-Well Plates, 210 Pc. 96-Well or 384-Well Plates, 150 Pc. Transwell or Insert Plates, 100 Pc. Deep-Well
Relative Humidity <95%
Temperature Range (Metric) 4°C to 50°C (with External Chiller)
Type Automated Incubator
Features Incubation and storage unit with temperature range below with external chiller, humidity up to 95% r.H. at 37°C and CO2 range of 0-20 Vol. % at 25°C to 50°C
Width (English) 31.5 in.
Width (Metric) 800 mm
Height (English) 36.0 in.
Height (Metric) 915 mm
Depth (English) 32.3 in.
Depth (Metric) 822 mm
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.