missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMM50 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06283PE
This item is not returnable.
View return policy
Description
SAMM50 Polyclonal antibody specifically detects SAMM50 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| SAMM50 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| CGI-51, FLJ42905, FLJ99036, OMP85, SAM50, sorting and assembly machinery 50kDa, sorting and assembly machinery component 50 homolog, sorting and assembly machinery component 50 homolog (S. cerevisiae), TOB55, Transformation-related gene 3 protein, transformation-related protein 3, TRG-3FLJ35825, YNL026W | |
| Produced in rabbits immunized with E. coli-derived Human SAMM50 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 25813 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction