missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gH Antibody (MSL-109) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43532
This item is not returnable.
View return policy
Description
Cytomegalovirus gH Monoclonal antibody specifically detects Cytomegalovirus gH in Cytomegalovirus samples. It is validated for Neutralization
Specifications
| Cytomegalovirus gH | |
| Monoclonal | |
| 1 mg/mL | |
| Neutralization | |
| envelope glycoprotein H, gH, gpUL75, HCMV gH, HHV-5 gH, HHV5 glycoprotein H, Human betaherpesvirus 5 envelope glycoprotein H, Human Cytomegalovirus glycoprotein H, UL75 | |
| CMV antigen (Virus antigen batch V2547) prepared from MRC-5 cells infected with the Towne strain of CMV. (Uniprot# Q4WHS2) | |
| 0.2 mg | |
| Primary | |
| Cytomegalovirus (CMV) | |
| Purified |
| Neutralization | |
| MSL-109 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3077450 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction