missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus p65 Antibody (rCMV/9827), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44629-100ug
This item is not returnable.
View return policy
Description
Cytomegalovirus p65 Monoclonal antibody specifically detects Cytomegalovirus p65 in Cytomegalovirus samples. It is validated for ELISA,Immunohistochemistry
Specifications
| Cytomegalovirus p65 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Cytomegalovirus (CMV) | |
| Purified |
| ELISA, Immunohistochemistry | |
| rCMV/9827 | |
| Unconjugated | |
| CMV, p65 | |
| Recombinant Cytomegalovirus p65 protein (Uniprot: P06725). Exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4⪚C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction