missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 1A1/SULT1A1 Rabbit anti-Human, Clone: 29, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00€ - 552.00€
Specifications
| Antigen | Cytosolic Sulfotransferase 1A1/SULT1A1 |
|---|---|
| Clone | 29 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283508
|
Novus Biologicals
NBP2-89949-50UL |
50 μg |
367.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18241208
|
Novus Biologicals
NBP2-89949-100UL |
100 μg |
552.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytosolic Sulfotransferase 1A1/SULT1A1 Monoclonal specifically detects Cytosolic Sulfotransferase 1A1/SULT1A1 in Human samples. It is validated for ELISA.Specifications
| Cytosolic Sulfotransferase 1A1/SULT1A1 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 6817 | |
| IgG | |
| Protein A purified |
| 29 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Aryl sulfotransferase 1, EC 2.8.2, EC 2.8.2.1, HAST1/HAST2, Phenol sulfotransferase 1, Phenol-sulfating phenol sulfotransferase 1, P-PST, P-PST 1, PST, ST1A1, ST1A3, STP1MGC5163, STPMGC131921, sulfotransferase 1A1, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1, Thermostable phenol sulfotransferase, thermostable phenol sulfotransferase1, ts-PST, TSPST1 | |
| Recombinant Human Cytosolic Sulfotransferase 1A1/SULT1A1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title