missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody (011), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89869AF350
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Monoclonal specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Sandwich ELISA Detection.
Specifications
| Cytosolic Sulfotransferase 1E1/SULT1E1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 11 | |
| Sandwich ELISA Detection | |
| EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic Sulfotransferase 1E1/SULT1E1 (NP_005411.1; Asn2-Ile294). | |
| 0.1 mL | |
| Breast Cancer, Stem Cell Markers | |
| 6783 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction