missing translation for 'onlineSavingsMsg'
Learn More

D 517 Hydrochloride (Verapamil Impurity), TRC

Product Code. 30358705
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358705 10 mg 10mg
30353170 50 mg 50mg
30359028 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358705 Supplier Toronto Research Chemicals Supplier No. TRCD10004510MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1794-55-4
Molecular Formula C26 H36 N2 O4 . Cl H
Molecular Weight (g/mol) 477.04
Synonym (2RS)-2-(3,4-Dimethoxyphenyl)-2-[2-[[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]ethyl]-3-methyl-butanenitrile Hydrochloride, Verapamil Hydrochloride Imp. I (EP) as Hydrochloride
IUPAC Name 2-(3,4-dimethoxyphenyl)-2-[2-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]ethyl]-3-methylbutanenitrile;hydrochloride
SMILES Cl.COc1ccc(CCN(C)CCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H36N2O4.ClH/c1-19(2)26(18-27,21-9-11-23(30-5)25(17-21)32-7)13-15-28(3)14-12-20-8-10-22(29-4)24(16-20)31-6;/h8-11,16-17,19H,12-15H2,1-7H3;1H
Formula Weight 476.2442
Percent Purity >95%
Chemical Name or Material (2RS)-2-(3,4-Dimethoxyphenyl)-2-[2-[[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]ethyl]-3-methyl-butanenitrile Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.