missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Amygdalin hydrate, 95%

Product Code. 10569440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10569440 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10569440 Supplier Thermo Scientific Acros Supplier No. 104730050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29883-15-6 | C20H27NO11 | 457.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29883-15-6
Melting Point 223.0°C to 226.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ELSD) (HPLC)
Packaging Glass bottle
Molecular Formula C20H27NO11
MDL Number MFCD29049822
Quantity 5 g
Beilstein 17, V,8, 118
Merck Index 15, 592
Specific Rotation − 38.50
Synonym amygdalin
Solubility Information (4% in water) Clear
InChI Key XUCIJNAGGSZNQT-ARGBPYSKNA-N
SMILES OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2R)-2-phenyl-2-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyacetonitrile
Molecular Weight (g/mol) 457.43
PubChem CID 66509032
Formula Weight 457.42
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(-)-Amygdalin hydrate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • D(-)-Amygdalin hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.