missing translation for 'onlineSavingsMsg'
Learn More

D-Arabinose, TRC

Product Code. 17957244
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17957244 50 g 50g
17959234 250 g 250g
17969314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17957244 Supplier Toronto Research Chemicals Supplier No. TRCA76417550G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28697-53-2
Molecular Formula C5 H10 O5
Molecular Weight (g/mol) 150.13
Synonym D-Arabinose, Arabinopyranose, D- (8CI), D-Arabinopyranose
IUPAC Name (3S,4R,5R)-oxane-2,3,4,5-tetrol
SMILES O[C@@H]1COC(O)[C@@H](O)[C@@H]1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3-,4+,5?/m1/s1
Formula Weight 150.0528
Chemical Name or Material D-Arabinopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.