Learn More
Invitrogen™ D-dimer Monoclonal Antibody (DD2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA120027
This item is not returnable.
View return policy
Description
MA1-20027 detects D-Dimer in human samples.
Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
Specifications
| D-dimer | |
| Monoclonal | |
| 6.8 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P02671 | |
| FGA | |
| High molecular weight fibrin degradation products or synthetic peptides covering the cross-linked region of D-dimer gamma-chain. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| DD2 | |
| Unconjugated | |
| FGA | |
| carboxy-terminal region; ENSMUSG00000059807; FGA; Fib; Fib2; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A; MGC119422; MGC119423; MGC119425 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2243 | |
| 4°C | |
| Liquid |