missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-N,N-Dimethylsphingosine, TRC

Product Code. 30179165
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179165 10 mg 10mg
30179573 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179165 Supplier Toronto Research Chemicals Supplier No. TRCD47925010MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119567-63-4
Molecular Formula C20 H41 N O2
Molecular Weight (g/mol) 327.54
IUPAC Name (E,2R,3S)-2-(dimethylamino)octadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](CO)N(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H41NO2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)19(18-22)21(2)3/h16-17,19-20,22-23H,4-15,18H2,1-3H3/b17-16+/t19-,20+/m1/s1
Formula Weight 327.3137
Chemical Name or Material D-erythro-N,N-Dimethylsphingosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.