missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Sphingosine, 99+%

Product Code. 15495779
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495779 10 mg 10mg
15405789 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495779 Supplier Thermo Scientific Alfa Aesar Supplier No. J63584.MA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-78-4 | C18H37NO2 | 299.499 g/mol

D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-78-4
Molecular Formula C18H37NO2
Molecular Weight (g/mol) 299.499
MDL Number MFCD00036751
InChI Key WWUZIQQURGPMPG-KRWOKUGFSA-N
Synonym sphingosine, d-erythro-sphingosine, 4-sphingenine, d-sphingosine, sphing-4-enine, cerebroside, sphingenine, 4-trans-sphingenine, trans-d-erythro-2-amino-4-octadecene-1,3-diol, 2s,3r,4e-2-amino-4-octadecene-1,3-diol
PubChem CID 5280335
ChEBI CHEBI:16393
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCCC=CC(C(CO)N)O

Specifications

Color White
Quantity 10 mg
Beilstein 1727294
Merck Index 14,8747
Solubility Information Soluble in ethanol.
Formula Weight 299.5
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D-erythro-Sphingosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.