Learn More
Alfa Aesar™ D-(+)-Fucose, 99%
Product Code. 11375159
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375159 5 g 5g
11365159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11375159 Supplier Alfa Aesar™ Supplier No. A18234.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Chemical Identifiers
CAS 3615-37-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00135603
InChI Key PNNNRSAQSRJVSB-DPYQTVNSSA-N
Synonym 2r,3s,4s,5r-2,3,4,5-tetrahydroxyhexanal, aldehydo-d-fucose, d +-fucose, galactose, 6-deoxy, d-galactose, 6-deoxy, fucose, d, +-fucose, d0o8ie, d-+-fucose, d-+-fucose;6-deoxy-d-galactose
PubChem CID 94270
ChEBI CHEBI:48203
IUPAC Name (2R,3S,4S,5R)-2,3,4,5-tetrahydroxyhexanal
SMILES CC(C(C(C(C=O)O)O)O)O
Specifications
Melting Point 135°C to 141°C
Quantity 5 g
Beilstein 1723320
Merck Index 14,4278
Optical Rotation +76° (c=10 in Water)
Formula Weight 164.16
Percent Purity 99%
Chemical Name or Material D-(+)-Fucose