missing translation for 'onlineSavingsMsg'
Learn More

D(+)Galactose Anhydrous, Extra Pure, SLR, Fisher Chemical

Product Code. 10437830 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10437830 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10437830 Supplier Fisher Chemical Supplier No. G/0050/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

100g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 59-23-4
Melting Point 165°C
Color White
pH 5.0 to 7
Packaging Plastic powder jar
Molecular Formula C6H12O6
MDL Number 151230
Quantity 100 g
Vapor Density 6.2
Formula Weight 180.16g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.