missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Galactose pentaacetate, 99+%

Product Code. p-137994 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646552 25 g Plastic bottle 25g
10357810 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10646552 Supplier Thermo Scientific Chemicals Supplier No. 158610250
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4163-60-4 | C16H22O11 | 390.341 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4163-60-4
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00063259
InChI Key LPTITAGPBXDDGR-LYYZXLFJSA-N
Synonym beta-d-galactose pentaacetate, penta-o-acetyl-beta-d-galactopyranose, b-d-galactose pentaacetate, beta-d-galactopyranose pentaacetate, beta-d-galactosepentaacetate, beta-d-galactopyranose, pentaacetate, 2s,3r,4s,5s,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4,6-penta-o-acetyl-beta-d-galactose, 1,2,3,4,6-penta-o-acetyl-beta-d-galactopyranose, 2r,3s,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate
PubChem CID 94752
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 25 g
Beilstein 31, 305
Specific Rotation Condition +25° (20°C c=10,CHC3,on dry ba)
Specific Rotation +24° to +26° (20°C, 589nm) (c=2, CHCl3)
Formula Weight 390.33
Percent Purity ≥99.0% (HPLC)
Physical Form Powder
Chemical Name or Material β-D-Galactose pentaacetate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.