missing translation for 'onlineSavingsMsg'
Learn More

D-Isoleucine, TRC

Product Code. 17989084
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989084 250 mg 250mg
17969084 50 mg 50mg
17939354 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17989084 Supplier Toronto Research Chemicals Supplier No. TRCI820210250MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319-78-8
Molecular Formula C6 H13 N O2
Molecular Weight (g/mol) 131.1729
Synonym Isoleucine, D- (8CI), (2R,3R)-2-Amino-3-methylpentanoic acid, (R)-Isoleucine
IUPAC Name (2R,3R)-2-amino-3-methylpentanoic acid
SMILES CC[C@@H](C)[C@@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m1/s1
Quantity 250 mg
Formula Weight 131.0946
Shipping Condition Room Temperature
Chemical Name or Material D-Isoleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.