missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D,L-m-Tyrosine, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | D,L-m-Tyrosine |
| CAS | 775-06-4 |
| Molecular Formula | C9 H11 N O3 |
| Quantity | 500 mg |
| Synonym | Phenylalanine','3-hydroxy-, DL-Phenylalanine','3-hydroxy-, m-Tyrosine','DL- (8CI), 3-Hydroxyphenylalanine, 2-Amino-3-(3-hydroxyphenyl)propanoic acid, 2-Azaniumyl-3-(3-hydroxyphenyl)propanoate, DL-3-(m-Hydroxyphenyl)alanine, DL-3-Hydroxyphenylalanine, DL-Metatyrosine, DL-m-Hydroxyphenylalanine, DL-m-Tyrosine, DL-α-m-Tyrosine, Metatyrosine, NSC 89304, m-DL-Tyrosine, m-Hydroxyphenylalanine, m-Tyrosine, meta-Tyrosine, α-Amino-3-hydroxybenzenepropanoic acid, α-Amino-β-(m-hydroxyphenyl)propionic acid |
| InChI Formula | InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-2-1-3-7(11)4-6/h1-4,8,11H,5,10H2,(H,12,13) |
| SMILES | NC(Cc1cccc(O)c1)C(=O)O |
| IUPAC Name | 2-amino-3-(3-hydroxyphenyl)propanoic acid |
| Molecular Weight (g/mol) | 181.19 |
| Formula Weight | 181.0739 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction