Learn More
D,L-O-Desmethyl Venlafaxine-d6, TRC
Especificaciones
Especificaciones
| Chemical Name or Material | D,L-O-Desmethyl Venlafaxine-d6 |
| CAS | 1020719-35-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16H19D6NO2 |
| Quantity | 1 mg |
| Synonym | 4-[2-(Dimethylamino)-1-(1-hydroxycyclo-hexyl)ethyl]phenol-d6 |
| InChI Formula | InChI=1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/i3D2,4D2,5D2 |
| SMILES | OC1(CC([2H])([2H])C([2H])([2H])C([2H])([2H])C1)C(CN(C)C)C2=CC=C(O)C=C2 |
| IUPAC Name | 3,3,4,4,5,5-hexadeuterio-1-[2-(dimethylamino)-1-(4-hydroxyphenyl)ethyl]cyclohexan-1-ol |
| Molecular Weight (g/mol) | 269.41 |
| Mostrar más |