missing translation for 'onlineSavingsMsg'
Learn More

D-Lactose Anhydrous, TRC

Product Code. 30007399
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007399 50 g 50g
30005043 5 g 5g
30003463 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007399 Supplier Toronto Research Chemicals Supplier No. TRCL11399550G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-42-3
Molecular Formula C12 H22 O11
Molecular Weight (g/mol) 342.3
Synonym Lactose, O-β-d-Galactopyranosyl-(1→4)-β-d-glucopyranose, D-Glucose, 4-O-β-D-galactopyranosyl-, Lactose (8CI), (+)-Lactose, AHL, Aletobiose, D-(+)-Lactose, DCL 15, DCl 11, Dairylac 80, Dilactose, Fast-Flo Lactose, Fast-flo, Flowlac 100, Galactinum, Granulac 140, Granulac 140M, Granulac 70, Inhalac 250, Inhalac 70, LE-PRO LAC 451, Lactin, Lactin (carbohydrate), Lactobiose, Lactohale 100, Lactohale 200, Lactohale 300, Lactose DCL 11, Lactose Edible 90, Lactose Fast-flo, Lactose anhydride, Lactose anhydrous, Milk sugar, Nonpareil 107, Osmolactan, Pharmatosa DCL 21, Pharmatose 21, Pharmatose 325M, Pharmatose DCL 15, PrenoLakt, Prismalac, Respitose MC 001, Respitose ML 001, Respitose ML 003, Respitose ML 006, Respitose SV 003, Respitose SV 010, Saccharum lactin, Sachelac, Sorbalac 400, Sorbolac 400, Spherolac, Super-Tab, SuperTab 11SD, SuperTab 21AN, Tablettose, Tablettose 70, Tablettose 80, Variolac 960, Zeparox EP
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5+,6+,7-,8-,9-,10-,11-,12+/m1/s1
Formula Weight 342.1162
Percent Purity >95%
Chemical Name or Material Lactose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.