missing translation for 'onlineSavingsMsg'
Learn More

Melford D-Luciferin Free Acid

Product Code. 30018432
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018432 50 mg 50mg
30018433 100 mg 100mg
30018434 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018432 Supplier Melford Supplier No. L370000.05
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used with firefly luciferase in bioluminescence for the determination of ATP.

Used with firefly luciferase in bioluminescence for the determination of ATP.

Specifications

CAS 2591-17-5
Molecular Formula C11H8N2O3S2
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.