missing translation for 'onlineSavingsMsg'
Learn More

D-Maltotriose, TRC

Product Code. 17938314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17938314 5 g 5g
17977354 25 g 25g
17907724 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938314 Supplier Toronto Research Chemicals Supplier No. TRCM1590005G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1109-28-0
Molecular Formula C18 H32 O16
Molecular Weight (g/mol) 504.44
Synonym Maltotriose (6CI,8CI), O-α-D-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose, Amylotriose, D-Maltotriose, NSC 170180, Triomaltose, Maltotriose (6CI,8CI), O-α-D-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose, Amylotriose, D-Maltotriose, NSC 170180, Triomaltose
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14-,15-,16?,17-,18-/m1/s1
Formula Weight 504.169
Percent Purity >95%
Chemical Name or Material D-(+)-Maltotriose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.