Learn More
D-Maltotriose, TRC
Product Code. 17977354
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17977354 25 g 25g
17938314 5 g 5g
17907724 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17977354 Supplier Toronto Research Chemicals Supplier No. TRCM15900025G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1109-28-0
Molecular Formula C18 H32 O16
Molecular Weight (g/mol) 504.44
Synonym Maltotriose (6CI,8CI), O-α-D-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose, Amylotriose, D-Maltotriose, NSC 170180, Triomaltose, Maltotriose (6CI,8CI), O-α-D-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose, Amylotriose, D-Maltotriose, NSC 170180, Triomaltose
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Specifications
Quantity 25 g
InChI Formula InChI=1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14-,15-,16?,17-,18-/m1/s1
Formula Weight 504.169
Percent Purity >95%
Chemical Name or Material D-(+)-Maltotriose
Grade HPLC