missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D14 Rabbit anti-Plant, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
250.00€ - 558.00€
Specifications
| Antigen | D14 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314516
|
Novus Biologicals
NBP3-16061-20UL |
20 μg |
250.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18350893
|
Bio-Techne
NBP3-16061-100UL |
100 μg |
558.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D14 Polyclonal antibody specifically detects D14 in Plant samples. It is validated for Western BlotSpecifications
| D14 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 4331983 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Plant | |
| D88, HTD2, LOC4331983, Protein DWARF 14, Protein DWARF 88, Protein HIGH-TILLERING DWARF 2, Strigolactone esterase D14, strigolactone esterase D14-like | |
| Recombinant protein of Oryza sativa D14. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title