missing translation for 'onlineSavingsMsg'
Learn More

D19 Reference Set (contains 2 of each of the following: D19-0, D19-100; and 1 of each of the following: D19-5, D19-10, D19-30, D19-50, D19-80, D19-120, D19-300, D19-500, D19-700, D19-900), VHG™

Product Code. 17907594
Change view
Click to view available options
Quantity:
1 gal
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17907594 1 gal 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17907594 Supplier VHG Labs Supplier No. VHG9150013551178

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

D19 Reference Set (contains 2 of each of the following: D19-0, D19-100; and 1 of each of the following: D19-5, D19-10, D19-30, D19-50, D19-80, D19-120, D19-300, D19-500, D19-700, D19-900)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Barium/Boron/Cadmium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
CAS 7429-90-5, 7440-39-3, 7440-42-8, 7440-43-9, 7440-47-3, 7440-50-8
Methods RDE
Analyte or Component Names Aluminium/Barium/Boron/Cadmium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
Certifications/Compliance ISO 17034
Solvent or Matrix Aviation Oil
Quantity 1 gal
Concentration or Composition (by Analyte or Components) Aluminium 0 μg/g/Barium 0 μg/g/Boron 0 μg/g/Cadmium 0 μg/g/Chromium 0 μg/g/Copper 0 μg/g/Iron 0 μg/g/Lead 0 μg/g/Magnesium 0 μg/g/Manganese 0 μg/g/Molybdenum 0 μg/g/Nickel 0 μg/g/Silver 0 μg/g/Sodium 0 μg/g/Tin 0 μg/g/Titanium 0 μg/g/Vanadium 0 μg/g/Zinc 0 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.