missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran Carboxamide Ethyl Ester, TRC

Product Code. 30132756
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132756 50 mg 50mg
17949959 5 mg 5mg
18043540 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132756 Supplier Toronto Research Chemicals Supplier No. TRCD10017050MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1422435-41-3
Molecular Formula C27 H28 N6 O4
Molecular Weight (g/mol) 500.55
Synonym Ethyl 3-[[2-[(4-carbamoylanilino)methyl]-1-methyl-benzimidazole-5-carbonyl]-(2-pyridyl)amino]propanoate, β-Alanine, N-[[2-[[[4-(aminocarbonyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-, ethyl ester
IUPAC Name ethyl 3-[[2-[(4-carbamoylanilino)methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate
SMILES CCOC(=O)CCN(C(=O)c1ccc2c(c1)nc(CNc3ccc(cc3)C(=O)N)n2C)c4ccccn4

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C27H28N6O4/c1-3-37-25(34)13-15-33(23-6-4-5-14-29-23)27(36)19-9-12-22-21(16-19)31-24(32(22)2)17-30-20-10-7-18(8-11-20)26(28)35/h4-12,14,16,30H,3,13,15,17H2,1-2H3,(H2,28,35)
Formula Weight 500.2172
Percent Purity >95%
Chemical Name or Material Dabigatran Carboxamide Ethyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.