missing translation for 'onlineSavingsMsg'
Learn More

Dalbavancin Hydrochloride, TRC

Product Code. 30133407
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133407 50 mg 50mg
30132708 250 mg 250mg
17993205 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133407 Supplier Toronto Research Chemicals Supplier No. TRCD11000050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Ristomycin A aglycone Hydrochloride, 5,31-dichloro-38-de(methoxycarbonyl)-7-demethyl-19-deoxy-56-O-[2-deoxy-2-[(10-methyl-1-oxoundecyl)amino]-beta-D-glucopyranuronosyl]-38-[[[3-(dimethylamino)propyl]amino]carbonyl]-42-O-alpha-D-mannopyranosyl-N15-methyl-∣5,31-Dichloro-38-de(methoxycarbonyl)-7-demethyl-19-deoxy-56-O-[2-deoxy-2-[(10-methyl-1-oxoundecyl)amino]-beta-D-glucopyranuronosyl]-38-[[[3-(dimethylamino)propyl]amino]carbonyl]-42-O-alpha-D-mannopyranosyl-N15-methylristomycin A aglycone∣A-A 1∣B 7011∣BI 397∣Dalbavancin∣Dalbavancin B0∣Dalvance∣MDL 63397∣VER 001

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 1853.1
Quantity 50 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.