missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daminozide-2,2,3,3-d4, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.01 g
0.05 g
missing translation for 'unitSize'
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Daminozide-d4 |
| Molecular Formula | C6 D4 H8 N2 O3 |
| Quantity | 0.01 g |
| Synonym | Daminozide D4 (Ethylene D4), Daminozide-d4, Butanedioic-2,2,3,3-d4 acid mono(2,2-dimethylhydrazide), Succinic-d4 N',N'-dimethylhydrazide, Succinic acid-d4 2,2-dimethylhydrazide |
| InChI Formula | InChI=1 S/C6H12N2O3/c1-8(2)7-5(9)3-4-6(10)11/h3-4H2,1-2H3,(H,7,9)(H,10,11)/i3D2,4D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C(=O)NN(C)C |
| IUPAC Name | 2,2,3,3-tetradeuterio-4-(2,2-dimethylhydrazinyl)-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 164.196 |
| Formula Weight | 164.11 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction