Learn More
Dansyl Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Dansyl Acid |
| CAS | 4272-77-9 |
| Molecular Formula | C12 H13 N O3 S |
| Quantity | 1 g |
| Synonym | 5-Dimethylaminonaphthalene-1-sulfonic Acid, 5-(Dimethylamino)-1-naphthalenesulfonic Acid |
| InChI Formula | InChI=1S/C12H13NO3S/c1-13(2)11-7-3-6-10-9(11)5-4-8-12(10)17(14,15)16/h3-8H,1-2H3,(H,14,15,16) |
| SMILES | CN(C)c1cccc2c(cccc12)S(=O)(=O)O |
| IUPAC Name | 5-(dimethylamino)naphthalene-1-sulfonic acid |
| Molecular Weight (g/mol) | 251.3 |
| Formula Weight | 251.0616 |