missing translation for 'onlineSavingsMsg'
Learn More

Dantrolene-13C3, TRC

Product Code. 30137721
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137721 10 mg 10mg
17960735 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137721 Supplier Toronto Research Chemicals Supplier No. TRCD18500210MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185234-99-4
Molecular Formula C1113C3H10N4O5
Molecular Weight (g/mol) 317.23
Synonym Dantrolene 13C3
IUPAC Name 1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione
SMILES [O-][N+](=O)c1ccc(cc1)c2oc(\C=N\N3[13CH2][13C](=O)N[13C]3=O)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H10N4O5/c19-13-8-17(14(20)16-13)15-7-11-5-6-12(23-11)9-1-3-10(4-2-9)18(21)22/h1-7H,8H2,(H,16,19,20)/b15-7+
Chemical Name or Material Dantrolene-13C3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.